Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate, 22 g/L, Reagents

Catalog No. 01335245
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Opaque HDPE
Catalog No. Quantity Packaging
01-335-245 1 L Glass Bottle
01-335-246 1 L Opaque HDPE
01-335-244 4 L Glass Bottle
3 options

Catalog No. 01-335-245

Supplier: Reagents Holdings Llc CF1313001C

Only null left
Add to Cart
Add to Cart

Ferrous Ammonium Sulfate, 22 g/L, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7783-85-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Iron(II) Ammonium Sulfate
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Light Green
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0178
Chemical Name or Material Ferrous Ammonium Sulfate, 22 g/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.