Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Ammonium Sulfate, Hexahydrate, Crystal, ACS, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7783-85-9
7783-85-9
Grade ACS
CAS Max % 1
Percent Purity 98.5 to 101.5%
Calcium (Ca) 5e-05
Copper (Cu) 3e-05
Insoluble Matter 0.0001
Magnesium (Mg) 2e-05
Manganese (Mn) 0.0001
Packaging Amber Glass Bottle
Phosphate 3e-05
Potassium (K) 2e-05
Sodium (Na) 0.0002
Zinc (Zn) 3e-05
Quantity 2.5 kg
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.