Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Fumarate, FCC, 97-101%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
141-01-5
C4H2FeO4
169.90
PMVSDNDAUGGCCE-ODZAUARKSA-L
λ²-iron(2+) (2Z)-but-2-enedioate
[Fe++].[O-]C(=O)\C=C/C([O-])=O
141-01-5
C4H2FeO4
169.90
PMVSDNDAUGGCCE-ODZAUARKSA-L
λ²-iron(2+) (2Z)-but-2-enedioate
[Fe++].[O-]C(=O)\C=C/C([O-])=O

F1054, 141-01-5, C4H2FeO4

Spectrum™ Ferrous Fumarate, FCC is used as an iron fortification food supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 97 to 101%
Lead (Pb) 2 mg/kg
Loss on Drying 0.015
Mercury (Hg) 3 mg/kg
Packaging Poly Pail
Sulfate 0.002
Quantity 12 kg