Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Gluconate, USP, 97-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
12389-15-0
C12H26FeO16
482.17
OKGNXSFAYMSVNN-UHFFFAOYNA-L
λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate
O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O
12389-15-0
C12H26FeO16
482.17
OKGNXSFAYMSVNN-UHFFFAOYNA-L
λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate
O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O

FE109, 12389-15-0, C12H22FeO14•2H2O

Spectrum™ Ferrous Gluconate, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 97 to 102%
Chloride 0.0007
Lead (Pb) 10 ppm
Loss on Drying 6.5-10%
Mercury (Hg) 3 ppm
Packaging Poly Pail
Sulfate 0.001
Quantity 12 kg