Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Lactate, Purified, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5905-52-2
5905-52-2
Grade Purified
CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.