Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Oxalate, Purified, 98%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6047-25-2
6047-25-2
Grade Purified
CAS Max % 1
Percent Purity 0.98
Chloride 5e-05
Lead (Pb) 5e-05
Packaging Fiber Drum
Quantity 25 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.