Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate, 7-Hydrate, Crystal, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
7782-63-0
FeH14O11S
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate,ferrous sulfate heptahydrate,presfersul,melanterite mineral,iron sulfate heptahydrate,iron 2+ sulfate heptahydrate,fesofor,fesotyme,haemofort,ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
7782-63-0
FeH14O11S
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate,ferrous sulfate heptahydrate,presfersul,melanterite mineral,iron sulfate heptahydrate,iron 2+ sulfate heptahydrate,fesofor,fesotyme,haemofort,ironate
62662
CHEBI:75836
iron(2+);sulfate;heptahydrate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
Formula Weight 278.02
Physical Form Crystals
Grade FCC/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.90kg
Quantity 2.5kg
Chemical Name or Material Ferrous Sulfate, 7-Hydrate