Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate, 7-Hydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7782-63-0
FeH14O11S
278.006
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
CHEBI:75836

High quality chemicals for laboratory and specialized industrial use

CAS 7782-63-0
Molecular Formula FeH14O11S
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
IUPAC Name iron(2+);sulfate;heptahydrate
Molecular Weight (g/mol) 278.006
PubChem CID 62662
ChEBI CHEBI:75836
Formula Weight 278.02
Physical Form Granular
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.90kg
Quantity 2.5kg
Chemical Name or Material Ferrous Sulfate, 7-Hydrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less