Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), Reagents

Catalog No. 01337958
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
25 kg

Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym Iron(II) Sulfate Heptahydrate
IUPAC Name λ²-iron(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Boiling Point >300°C
Color Green or Bluish-Green
Physical Form Solid
Quantity 25 kg
Grade ACS
Packaging Fiber Drum
Specific Gravity 1.897
Chemical Name or Material Ferrous Sulfate Heptahydrate, ACS Reagent (Iron (II) Sulfate Heptahydrate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.