Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7782-63-0
FeH14O11S
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
7782-63-0
FeH14O11S
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

F1060, 7782-63-0, FeSO4•7H2O

Spectrum™ Ferrous Sulfate, Heptahydrate, Crystal, Reagent, ACS As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
CAS Max % 1
Percent Purity 99%
Calcium (Ca) 5e-005
Chloride 1e-005
Copper (Cu) 5e-005
Insoluble Matter 0.0001
Magnesium (Mg) 2e-005
Manganese (Mn) 0.0005
Packaging Poly Pail
Phosphate 1e-005
Potassium (K) 2e-005
Sodium (Na) 0.0002
Zinc (Zn) 5e-005
Quantity 12 kg
Show More Show Less