Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferrous Sulfate, Heptahydrate, Granular, USP, 99.5-104.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7782-63-0
FeH14O11S
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
7782-63-0
FeH14O11S
278.01
SURQXAFEQWPFPV-UHFFFAOYSA-L
λ²-iron(2+) heptahydrate sulfate
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

FE110, 7782-63-0, FeSO4•7H2O

Spectrum™ Ferrous Sulfate, Heptahydrate, Granular, USP is used to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 99.5 to 104.5%
Lead (Pb) 10 ppm
Mercury (Hg) 3 μg/g
Packaging Poly Pail
Quantity 2.5 kg