Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferulic Acid, Powder, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
20 kg
Packaging:
Amber Glass Bottle
Fiber Drum
1135-24-6
1135-24-6
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.