Learn More
Omega Scientific Inc Fetal Bovine Serum, Charcoal-Stripped/ Dextran-Treated, 500 mL
Supplier:  Omega Scientific Inc FB04
Encompass_Preferred
Charcoal-Stripped/ Dextran-Treated FBS has lower levels of hormones, steroids, and growth factors which are reduced using charcoal, dextran and NaCl until progesterone concentration is less than 0.2 ng/mL. A hormone panel is performed pre and post-treatment and reported on the COA. 500 mL
Catalog No. NC2117586
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.