Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Australia Available on GSA/VA Contract for Federal Government customers only.
Standard sera for robust cell lines
$1068.00
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
10-100-147 Greener Choice Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Gibco™
10100147
500mL
Each for $1,068.00
Add to cart
Description
Description
  • Sera Category: Secure
  • Origin: Australia
  • Endotoxin level: ≤10EU/mL
  • Hemoglobin level: ≤30mg/dL (levels routinely ≤25mg/dL)
Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.