Learn More
Fetal Hemoglobin Mouse anti-Human, PE, Clone: HBF-1, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHFH044
 View more versions of this product
Catalog No. 50-112-1474
Add to cart
This item is not returnable. View return policy
Description
Description
The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5 ft.- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3 ft.. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5 ft. untranslated regions and the introns, but they differ significantly over the 3 ft. untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
Specifications
Specifications
Fetal Hemoglobin
Monoclonal
PE
HBG1
Liquid
HBG1
Human Fetal Hemoglobin (HbF)
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HBF-1
PBS with <10% sucrose, BSA and 0.1% sodium azide
P69891
alpha-1 globin, hemoglobin alpha 1, hemoglobin subunit alpha, Hemoglobin subunit gamma-1, HBG1, Gamma-1-globin, Hb F Agamma, Hemoglobin gamma-1 chain, Hemoglobin gamma-A chain
Mouse
IgG1
2 mL
4° C, store in dark
Primary
3047