Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ α-Fetoprotein (AFP) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ α-Fetoprotein (AFP) Ab-1, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS226P1ABX

 View more versions of this product

Catalog No. MS226P1ABX

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Host Species: Mouse

Clone: C3

Isotype: IgG2a

Species Reactivity: Human, Dog and Pig. It does not react with cow, cat, mouse and rat.

Epitope: Not determined

Immunogen: Raised against affinity purified α-fetoprotein (AFP) from serum of a hepatoma patient

Molecular Weight: 70 kDa

Positive Control: Fetal liver or hepatocellular CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

alpha-Fetoprotein (AFP) Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Oncogenesis
Canine, Human, Porcine
IgG2a
Immunohistochemistry (Paraffin)
C3
Unconjugated
Raised against affinity purified alpha fetoprotein (AFP) from serum of a hepatoma patient
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit