Learn More
Epredia™ Lab Vision™ α-Fetoprotein (AFP) Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ α-Fetoprotein (AFP) Ab-1, Mouse Monoclonal Antibody.
Specifications
Antigen | alpha-Fetoprotein (AFP) Ab-1 |
---|---|
Clone | C3 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Concentration | Quantity | Price | Quantity & Availability | ||||
MS226P1
|
Epredia™
MS226P1 |
200μg/mL | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS226P1ABX
|
Epredia™
MS226P1ABX |
1mg/mL | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
MS226PABX
|
Epredia™
MS226PABX |
1mg/mL | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.Host Species: Mouse
Clone: C3
Isotype: IgG2a
Species Reactivity: Human, Dog and Pig. It does not react with cow, cat, mouse and rat.
Epitope: Not determined
Immunogen: Raised against affinity purified α-fetoprotein (AFP) from serum of a hepatoma patient
Molecular Weight: 70 kDa
Positive Control: Fetal liver or hepatocellular CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry
Note:
USA: RUO; Int'l: RUO
Specifications
alpha-Fetoprotein (AFP) Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Oncogenesis | |
Raised against affinity purified alpha fetoprotein (AFP) from serum of a hepatoma patient | |
Primary |
C3 | |
Monoclonal | |
Concentrated | |
RUO | |
Canine, Human, Porcine | |
IgG2a |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.