Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody.

$241.50 - $386.00

Specifications

Antigen alpha Fetoprotein (AFP)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 6
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RB9064P
View Documents
Thermo Scientific™
RB9064P
Purified Ab with BSA and Azide 1mL Each for $385.50
Add to cart
 
RB9064P1
View Documents
Thermo Scientific™
RB9064P1
Purified Ab with BSA and Azide 500μL Each for $241.50
Add to cart
 
RB9064R7
View Documents
Thermo Scientific™
RB9064R7
Ready-to-use for IHC (UVLP) 7mL Each for $324.00
Add to cart
 
RB9064RQ
View Documents
Thermo Scientific™
RB9064RQ
Ready-to-use for IHC (Quanto) 12mL Each for $386.00
Add to cart
 
RB9064P0
View Documents
Thermo Scientific™
RB9064P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9064R1
View Documents
Thermo Scientific™
RB9064R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody.

AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Human α-fetoprotein purified from human cord serum

Molecular Weight: 70 kDa

Positive Control: HeLa cells, HepG2 cells, fetal liver or hepatocellular CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

alpha Fetoprotein (AFP)
Polyclonal
Rabbit
Oncogenesis
Human alpha-fetoprotein purified from human cord serum
Western Blot
Unconjugated
IVD
Primary
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit