Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ α-Fetoprotein (AFP), Rabbit Polyclonal Antibody.

Supplier:  Epredia™ RB9064R1

 View more versions of this product

Catalog No. RB9064R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Human α-fetoprotein purified from human cord serum

Molecular Weight: 70 kDa

Positive Control: HeLa cells, HepG2 cells, fetal liver or hepatocellular CA

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

alpha Fetoprotein (AFP)
Polyclonal
Unconjugated
Rabbit
1 mL
Oncogenesis
Human
IgG
Immunohistochemistry (Paraffin), Western Blot
Polyclonal
Ready-to-use for IHC (UVLP)
Human alpha-fetoprotein purified from human cord serum
IVD
Primary
Prediluted
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit