Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEV Polyclonal Antibody
FEV Polyclonal Antibody
Supplier: Thermo Scientific PA541153
Description
FEV Polyclonal Antibody for Western Blot

Specifications
FEV | |
Polyclonal | |
Unconjugated | |
FEV | |
ETS domain transcription factor Pet-1; ETS-domain transcription factor Pet-1; Fev; FEV (ETS oncogene family); FEV (fifth Ewing variant); FEV transcription factor, ETS family member; FEV, ETS transcription factor; fifth Ewing variant protein; HSRNAFEV; mPet-1; PC12 ETS domain-containing transcription factor 1; PC12 ETS factor 1; Pet1; Pet-1; pheochromocytoma 12 ETS (E26 transformation-specific); protein FEV | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54738 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q99581 | |
FEV | |
Synthetic peptide directed towards the C-terminal of human FEV (aa 149-198). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction