Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Fexofenadine hydrochloride

Click to view available options
Quantity:
25mg
100mg
500mg
153439-40-8
C32H40ClNO4
538.125
MFCD00865710
RRJFVPUCXDGFJB-UHFFFAOYSA-N
fexofenadine hydrochloride,fexofenadine hcl,allegra,telfast,2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride,fexofenidine hydrochloride,fexofenadine hydrochloride,terfenadine carboxylate hydrochloride,mdl 16455 hydrochloride,terfenidine carboxylate hydrochloride
63002
2-[4-[1-hydroxy-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butyl]phenyl]-2-methylpropanoic acid;hydrochloride
CC(C)(C1=CC=C(C=C1)C(CCCN2CCC(CC2)C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O)C(=O)O.Cl
153439-40-8
C32H40ClNO4
538.125
MFCD00865710
RRJFVPUCXDGFJB-UHFFFAOYSA-N
fexofenadine hydrochloride,fexofenadine hcl,allegra,telfast,2-4-1-hydroxy-4-4-hydroxydiphenylmethyl piperidin-1-yl butyl phenyl-2-methylpropanoic acid hydrochloride,fexofenidine hydrochloride,fexofenadine hydrochloride,terfenadine carboxylate hydrochloride,mdl 16455 hydrochloride,terfenidine carboxylate hydrochloride
63002
2-[4-[1-hydroxy-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butyl]phenyl]-2-methylpropanoic acid;hydrochloride
CC(C)(C1=CC=C(C=C1)C(CCCN2CCC(CC2)C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O)C(=O)O.Cl

H-1 receptor antagonist

Applications
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.

Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
Color White
Melting Point 148°C to 150°C
Quantity 500mg
Merck Index 14,4068
Formula Weight 538.12
Physical Form Crystalline Powder
Chemical Name or Material Fexofenadine hydrochloride

RTECSNumber : CY1633377

Recommended Storage : −30° to −10°C