Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fexofenadine hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Fexofenadine hydrochloride is a selective histamine H1 receptor antagonist (pKi = 8.1). Active metabolite of terfenadine that displays non-sedating antiallergic effects. Fexofenadine hydrochloride counteracts histamnine mediated immune suppression and improves immunotherapy response.
Specifications
Quantity | 50 mg |
Formula Weight | 538.12 |
Percent Purity | >99% |
Chemical Name or Material | Fexofenadine hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction