Fexofenadine hydrochloride, Tocris

H1 receptor antagonist; non-sedating antiallergic agent

Manufacturer: R&D Systems 2429/10

Quantity 10mg

 View more versions of this product

Catalog No. 24-291-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C32H39NO4·HCl
    Formula Weight 538.12
    Synonym α, α-Dimethyl-4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]butyl]-benzeneacetic acid hydrochloride
    Chemical Name or Material Fexofenadine hydrochloride

    Specifications

    Formula Weight 538.12
    Quantity 10mg
    Molecular Formula C32H39NO4·HCl
    Synonym α, α-Dimethyl-4-[1-hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]butyl]-benzeneacetic acid hydrochloride
    Recommended Storage Desiccate at +4°C
    Assay Percent Range >99%
    CAS 153439-40-8
    Chemical Name or Material Fexofenadine hydrochloride

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit