Learn More
Aviva Systems Biology Corp FEZF1 Antibody - C-terminal region (ARP50396_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FEZF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FEZF1. Alternate names: ZNF312B, FEZApplicationIHC, WB. Species reactivity: Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-60335