Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FEZF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543562
Description
Peptide sequence: CPTCGKGFCR NFDLKKHVRK LHDSSLGLAR TPAGEPGTEP PPPLPQQPPM Sequence homology: Guinea Pig: 79%; Horse: 79%; Human: 100%; Rabbit: 92%; Rat: 100%.
Transcription repressor. Involved in the axonal projection and proper termination of olfactory sensory neurons (OSN). Plays a role in rostro-caudal patterning of the diencephalon and in prethalamic formation. Expression is required in OSN to cell-autonomously regulate OSN axon projections. Regulates non-cell-autonomously the layer formation of the olfactory bulb development and the interneurons. May be required for correct rostral migration of the interneuron progenitors.
Specifications
FEZF1 | |
Polyclonal | |
Unconjugated | |
FEZF1 | |
3110069A13Rik; FEZ; Fez family zinc finger 1; fez family zinc finger protein 1; Fezf1; fez-like; HH22; LOW QUALITY PROTEIN: fez family zinc finger protein 1; RGD1560480; Zfp312-like; zinc finger protein 312B; ZNF312B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
389549 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A0PJY2 | |
FEZF1 | |
Synthetic peptide directed towards the C-terminal of human FEZF1 (aa 402-451). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction