Learn More
FFAR2/GPR43 Mouse anti-Human, Alexa Fluor™ 750, Clone: 1000624, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB10082S100UG
Catalog No. FB10082S100
Add to cart
This item is not returnable. View return policy
Description
Description
FFAR2/GPR43 Monoclonal antibody specifically detects FFAR2/GPR43 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
FFAR2/GPR43
1000624
Flow Cytometry 0.25-1 μg/106 cells
O15552
free fatty acid activated receptor 2, free fatty acid receptor 2, G protein-coupled receptor 43, GPR43, GPR43FFA2R, G-protein coupled receptor 43
Mouse myeloma cell line NS0-derived recombinant human FFAR2/GPR43, Met1-Glu330, Accession # O15552
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human FFAR2/GPR43 in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
2867
Human