Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ GeneAmp™ dNTP Blend (100 mM) Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Dissolved in glass-distilled water and titrated to pH 7.0 with NaOH

Manufacturer:  Applied Biosystems™ N8080261

 View more versions of this product

Catalog No. N8080261


Add to cart

Description

Description

GeneAmp dNTPs are deoxynucleotide triphosphates wiht high purity,making them ideal for use in PCR reactions or other applications.

  • Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
  • Premixed dNTP blends are available in various concentrations, offering ease of use
  • Each vial contains blend of 100mM solution of dNTP (25mM each of dATP, dCTP, dGTP and dTTP)
  • Sufficient for 2000 PCR amplifications, 50µL each
  • Kit contains 12 x 1mL vials

PCR, PCR and Real-Time PCR

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

100 mM
Store at -20°C in a constant temperature freezer.
GeneAmp®
1mL
Dry Ice
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.