Learn More
FGD5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209101AP
Catalog No. 50-173-3150
Add to cart
Description
Description
FGD5 activates CDC42, a member of the Ras-like family of Rho- and Rac proteins, by exchanging bound GDP for free GTP. It mediates VEGF-induced CDC42 activation. It may regulate proangiogenic action of VEGF in vascular endothelial cells, including network formation, directional movement and proliferation and may play a role in regulating the actin cytoskeleton and cell shape.
Specifications
Specifications
FGD5
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZNL6, Q80UZ0
FGD5, ZFYVE23
Rabbit
IgG
150 μL
-20°C
Primary
152273, 232237, 362402
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
FGD5
Liquid
Fgd5
FGD5 Fusion Protein Ag15037
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat