Learn More
FGF-11 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0282-6J20, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212296AF594
Catalog No. NP212296A94
Add to cart
Description
Description
FGF-11 Monoclonal antibody specifically detects FGF-11 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FGF-11
MM0282-6J20
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FGF-11, FHF-3, FHF3Fibroblast growth factor homologous factor 3, fibroblast growth factor 11, FLJ16061, MGC102953, MGC45269
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
FGF11
Human FGF11
0.1 mL
Primary
2256