Learn More
FGF-13 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S235-22, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242178V
Catalog No. NBP242178V
Add to cart
Description
Description
FGF-13 Monoclonal antibody specifically detects FGF-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
FGF-13
S235-22
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
FGF13
Synthetic peptide amino acids 2-18 (AAAIASSLIRQKRQARE) of human FHF2A. 100% identical to rat, 94% identical to mouse. >80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A.
Protein G purified
Angiogenesis, Biologically Active Proteins, Neuroscience
Primary
2258
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Greater then 80% identity with FGF12A/FHF1A, FGF14A/FHF4A and FGF11A/FHF3A. Detects approx 30kDa. Does not cross-react with FGF13B/FHF2B. Cross reacts with FGF12A/FHF1A and FGF14A/FHF4A.