Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-13 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI7E8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270721JF549

Catalog No. N270721J549


Add to cart

Description

Description

FGF-13 Monoclonal antibody specifically detects FGF-13 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

FGF-13
OTI7E8
Immunohistochemistry
Affinity Purified, Conjugated
FGF13
Full length recombinant protein of human FGF-13 (NP_004105) produced in E.coli.
Immunogen affinity purified
Angiogenesis, Biologically Active Proteins, Neuroscience
Primary
2258.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
FGF-13, FGF2Fibroblast growth factor homologous factor 2, FHF2FHF-2, fibroblast growth factor 13
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit