Learn More
FGF-16 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0283-6G2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212297AF594
Catalog No. NP212297A94
Add to cart
Description
Description
FGF-16 Monoclonal antibody specifically detects FGF-16 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FGF-16
MM0283-6G2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FGF-16, fibroblast growth factor 16
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
FGF16
Human FGF16
0.1 mL
Primary
8823