Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP309433100UL

 View more versions of this product

Catalog No. NB123767


Add to cart

Description

Description

FGF-16 Polyclonal specifically detects FGF-16 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FGF-16
Unconjugated
PBS buffer, 2% sucrose
FGF-16, fibroblast growth factor 16
The immunogen is a synthetic peptide directed towards the C-terminal region of Human FGF-16 (NP_003859). Peptide sequence REQFEENWYNTYASTLYKHSDSERQYYVALNKDGSPREGYRTKRHQKFTH
100 μg
Angiogenesis
Primary
8823
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit