Learn More
FGF-16 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF1212SP
Catalog No. AF1212SP
Add to cart
This item is not returnable. View return policy
Description
Description
FGF-16 Polyclonal specifically detects FGF-16 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
Specifications
FGF-16
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Neutralization 3-9 ug/mL, Immunocytochemistry 5-15 ug/mL
O43320
FGF16, FGF-16, fibroblast growth factor 16
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human FGF-16 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-3, -4, -5, -6, -7, -9, -10, -11, -12, -13, -17, -18, -19, -21, -23, recombinant mouse (rm) FGF-8b, and rmFGF-15 is observed.
Human, Mouse
Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
FGF16
E. coli-derived recombinant human FGF-16 Ala2-Arg207 Accession # O43320
Antigen Affinity-purified
RUO
Primary
8823
Reconstitute at 0.2 mg/mL in sterile PBS.