Learn More
FGF-17 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0284-5H27, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212298AF532
Catalog No. NP212298A32
Add to cart
Description
Description
FGF-17 Monoclonal antibody specifically detects FGF-17 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
FGF-17
MM0284-5H27
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
FGF-13, FGF-17, fibroblast growth factor 17
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FGF17
Human FGF17
0.1 mL
Primary
8822