Learn More
FGF-19 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0285-4G16, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212299AF750
Catalog No. NP212299A70
Add to cart
Description
Description
FGF-19 Monoclonal antibody specifically detects FGF-19 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FGF-19
MM0285-4G16
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FGF-19, fibroblast growth factor 19
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
FGF19
Human FGF19
0.1 mL
Primary
9965