Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-21 Mouse anti-Human, PE/Atto594, Clone: OTI2F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271815PEATT594

Catalog No. N271815P594


Add to cart

Description

Description

FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FGF-21
OTI2F10
Flow Cytometry
Affinity Purified, Conjugated
FGF21
Recombinant protein fragment corresponding to amino acids 29-209 of human FGF-21 (NP_061986) produced in E.coli.
Immunogen affinity purified
Angiogenesis, Biologically Active Proteins, Cancer
Primary
26291.0
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
FGF-21, fibroblast growth factor 21
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit