Learn More
FGF-3 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0287-3F18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212301AF750
Catalog No. NP212301A70
Add to cart
Description
Description
FGF-3 Monoclonal antibody specifically detects FGF-3 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FGF-3
MM0287-3F18
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
fibroblast growth factor 3, fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog), HBGF-3, mouse, oncogene INT2
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
FGF3
Human FGF-3
0.1 mL
Primary
2248