Learn More
FGF-4 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0288-9H51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212302AF532
Catalog No. NP212302A32
Add to cart
Description
Description
FGF-4 Monoclonal antibody specifically detects FGF-4 in Human samples. It is validated for Western Blot.
Specifications
Specifications
FGF-4
MM0288-9H51
Western Blot
fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FGF4
Human FGF4
0.1 mL
Primary
2249