Learn More
FGF-4 Mouse anti-Human, DyLight 755, Clone: 3A12G9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237374IR
Catalog No. NBP237374IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
FGF-4
3A12G9
Western Blot, ELISA
Purified
FGF4
Purified recombinant fragment of humanFGF4 (AA: 62-123) expressed in E. Coli.
Protein A or G purified
Angiogenesis, Biologically Active Proteins, Cancer
Primary
2249
ELISA, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
fibroblast growth factor 4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human