Learn More
FGF-5 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0289-8M27, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212303AF532
Catalog No. NP212303A32
Add to cart
Description
Description
FGF-5 Monoclonal antibody specifically detects FGF-5 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FGF-5
MM0289-8M27
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FGF-5, fibroblast growth factor 5, HBGF-5, Heparin-binding growth factor 5, Smag-82
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FGF5
Human FGF5
0.1 mL
Primary
2250