Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGF-6 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF238SP
This item is not returnable.
View return policy
Description
FGF-6 Polyclonal specifically detects FGF-6 in Human samples. It is validated for Western Blot, Neutralization.Specifications
FGF-6 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FGF6, FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2 | |
Goat | |
Affinity Purified | |
RUO | |
2251 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.2-1 ug/mL | |
P10767 | |
FGF6 | |
E. coli-derived recombinant human FGF-6 Gly41-Ile208 Accession # P10767 | |
25 μg | |
Primary | |
Detects human FGF-6 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human (rh) FGF-4 is observed, less than 5% cross-reactivity with rhFGF-9 is observed, and less than 2% cross-reactivity with FGF-acidic, rhFGF-5, and rhFGF-7 is observed. In direct ELISAs, less than 1% cross-reactivity with rh beta-ECGF, recombinant mouse (rm) FGF-8b, and rmFGF-8c is observed. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction