Learn More
FGF-6 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0290-8N21, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212304AF532
Catalog No. NP212304A32
Add to cart
Description
Description
FGF-6 Monoclonal antibody specifically detects FGF-6 in Human samples. It is validated for ELISA.
Specifications
Specifications
FGF-6
MM0290-8N21
ELISA
FGF-6, fibroblast growth factor 6, HBGF-6, Heparin secretory-transforming protein 2, Heparin-binding growth factor 6, HST2, HST-2, HSTF2, HSTF-2
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
FGF6
Human FGF6
0.1 mL
Primary
2251