Learn More
FGF-9 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0292-4D25, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212306AF594
Catalog No. NP212306A94
Add to cart
Description
Description
FGF-9 Monoclonal antibody specifically detects FGF-9 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
FGF-9
MM0292-4D25
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
FGF9
Human FGF9
0.1 mL
Primary
2254