Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF acidic/FGF1 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI4D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270717JF549

Catalog No. N270717J549


Add to cart

Description

Description

FGF acidic/FGF1 Monoclonal antibody specifically detects FGF acidic/FGF1 in Human samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

FGF acidic/FGF1
OTI4D2
Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
FGF1
Recombinant protein fragment corresponding to amino acids 16-155 of human FGF acidic/FGF1 (NP_000791) produced in E.coli.
Immunogen affinity purified
Angiogenesis, Biologically Active Proteins, Cancer, Stem Cell Markers
Primary
2246.0
Immunocytochemistry, Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit