Learn More
FGF-BP Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0293-5K23, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212307AF350
Catalog No. NP212307A30
Add to cart
Description
Description
FGF-BP Monoclonal antibody specifically detects FGF-BP in Human samples. It is validated for Western Blot.
Specifications
Specifications
FGF-BP
MM0293-5K23
Western Blot
17 kDa heparin-binding growth factor-binding protein, FGF-BP, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FGFBP1
Human FGFBP1
0.1 mL
Primary
9982