Learn More
FGF-BP Mouse anti-Human, Alexa Fluor 488, Clone: MM0293-5K23, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212307AF488
Catalog No. NB212307488
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
FGF-BP Monoclonal antibody specifically detects FGF-BP in Human samples. It is validated for Western Blot.
Specifications
Specifications
FGF-BP
MM0293-5K23
Western Blot
Purified
FGFBP1
Human FGFBP1
Protein G purified
Angiogenesis, Cell Cycle and Replication
Primary
9982
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
17 kDa heparin-binding growth factor-binding protein, FGF-BP, FGF-BP1, FGFBP-1, FGFBPFGF-binding protein 1, fibroblast growth factor binding protein 1, fibroblast growth factor-binding protein 1, HBp17, HBP1717 kDa HBGF-binding protein, heparin-binding growth factor binding protein
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human