Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR2 Mouse anti-Human, Alexa Fluor 350, Clone: 98725, R&D Systems

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB684U100UG

Catalog No. FAB684U100


Add to cart
This item is not returnable. View return policy

Description

Description

FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

FGFR2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Mouse
IgG1
RUO
Primary
2263
Human
Flow Cytometry
98725
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FGFR2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects alpha isoforms (3 Ig-like domains) and beta isoforms (2 Ig-like domains) of human FGF R2 in direct ELISAs and Western blots. In direct ELISAs, this antibody reacts with all isoforms of rhFGF R2 but shows approximately a 4-fold preference for (IIIc) isoforms. Approximately 100% cross-reactivity with recombinant mouse FGF R2, 15% cross-reactivity with recombinant human (rh) FGF RI ( beta isoforms), and no cross-reactivity with rhFGF RI ( alpha isoforms), rhFGF R3, or rhFGF R4 is observed.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)