Learn More
FGFR3 Mouse anti-Human, Alexa Fluor 750, Clone: 136334, R&D Systems
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB766S100UG
Catalog No. FAB766S100
Add to cart
This item is not returnable. View return policy
Description
Description
FGFR3 Monoclonal antibody specifically detects FGFR3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
FGFR3
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism
Mouse
IgG1
RUO
Primary
2261
Human
Flow Cytometry
136334
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
FGFR3
Mouse myeloma cell line NS0- and Sf21-derived recombinant human FGF R3 alpha (IIIb)
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects the IIIb and IIIc isoforms of human FGF R3 in direct ELISAs and Western blots. Does not cross-react with any isoforms of recombinant mouse (rm) FGF R3, rmFGF R2, recombinant human (rh) FGF R1, rhFGF R2, or rhFGF R4.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)