Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FGF R3 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene Alias EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism
Gene ID (Entrez) 2261
Gene Symbol FGFR3
Name Recombinant FGF R3 Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60208250U View Documents Novus Biologicals™
NBP260208250UG
250μg Each for $1,025.00
Add to cart
 
NB6020850UG View Documents Novus Biologicals™
NBP26020850UG
50μg Each for $287.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal hIgG-His-tag and corresponding to the amino acids 23-375 of Human FGF R3 The Recombinant Human FGFR3 Protein is derived from Baculovirus. The Recombinant Human FGFR3 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 10% glycerol. containingno Preservative
2261
Recombinant FGF R3 Protein
Baculovirus
1mg/mL
SDS-Page
EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism
FGFR3
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit